siRNA Details
| Gene Symbol: SPG20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045180.1 | 4 | 353 | |||
| NR_045181.1 | 3 | 266 | ||||
| GenBank mRNA | AW292824.1 | - | 4 | 352 | ||
| BF732820.1 | - | 3 | 266 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPG20-AS1| Gene Symbol: SPG20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045180.1 | 4 | 353 | |||
| NR_045181.1 | 3 | 266 | ||||
| GenBank mRNA | AW292824.1 | - | 4 | 352 | ||
| BF732820.1 | - | 3 | 266 | |||