siRNA Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005248333.4 | XP_005248390.1 | 1 | 523 | ||
| XM_006714486.3 | XP_006714549.1 | 1 | 523 | |||
| XM_011514074.2 | XP_011512376.1 | 1 | 523 | |||
| XM_011514075.2 | XP_011512377.1 | 1 | 523 | |||

TMEM267
Genomic Map