siRNA Details
| Gene Symbol: FAM188B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007621.1 | XP_016863110.1 | 1 | 563 | ||
| GenBank mRNA | HM626137.1 | - | 7 | 895 | ||
| HM626138.1 | - | 3 | 473 | |||
| HM626139.1 | - | 4 | 460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM188B2| Gene Symbol: FAM188B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007621.1 | XP_016863110.1 | 1 | 563 | ||
| GenBank mRNA | HM626137.1 | - | 7 | 895 | ||
| HM626138.1 | - | 3 | 473 | |||
| HM626139.1 | - | 4 | 460 | |||