siRNA Details
Gene Symbol: FAM188B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007621.1 | XP_016863110.1 | 5 | 944 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM188B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007621.1 | XP_016863110.1 | 5 | 944 |