siRNA Details
| Gene Symbol: PRSS56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195129.1 | NP_001182058.1 | 8 | 1037 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS56| Gene Symbol: PRSS56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195129.1 | NP_001182058.1 | 8 | 1037 | ||