siRNA Details
| Gene Symbol: HRCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039792.1 | NP_001034881.1 | 1 | 922 | ||
| GenBank mRNA | AY358408.1 | - | 1 | 922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HRCT1| Gene Symbol: HRCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039792.1 | NP_001034881.1 | 1 | 922 | ||
| GenBank mRNA | AY358408.1 | - | 1 | 922 | ||