siRNA Details
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146686.2 | NP_001140158.1 | 5 | 799 | ||
| GenBank mRNA | BC031680.1 | - | 1 | 544 | ||
| CR749823.1 | - | 2 | 632 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GMNC| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146686.2 | NP_001140158.1 | 5 | 799 | ||
| GenBank mRNA | BC031680.1 | - | 1 | 544 | ||
| CR749823.1 | - | 2 | 632 | |||