siRNA Details
| Gene Symbol: LOC105369140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003053.1 | XP_016858542.1 | 1, 2 | 712 | ||
| XM_017003054.1 | XP_016858543.1 | 1, 2 | 712 | |||
| XM_017003055.1 | XP_016858544.1 | 1, 2 | 712 | |||

LOC105369140
Genomic Map