siRNA Details
| Gene Symbol: GDF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004962.3 | NP_004953.1 | 2 | 1606 | ||
| GenBank mRNA | AK310360.1 | - | 2 | 1463 | ||
| AK315322.1 | - | 2 | 1459 | |||
| BC028237.1 | - | 2 | 1036 | 5245100 | ||
| D49492.1 | - | 2 | 1444 | |||
| L42113.1 | - | 1 | 101 | |||

GDF10
Genomic Map