siRNA Details
| Gene Symbol: GDF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004962.3 | NP_004953.1 | 2, 3 | 1680 | ||
| GenBank mRNA | AK315322.1 | - | 2, 3 | 1533 | ||
| BC028237.1 | - | 2, 3 | 1110 | 5245100 | ||
| D49492.1 | - | 2, 3 | 1518 | |||
| L42113.1 | - | 1, 2 | 175 | |||

GDF10
Genomic Map