siRNA Details
| Gene Symbol: PRAMENP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135291.1 | 6 | 2070 | |||
| GenBank mRNA | AK131325.1 | - | 6 | 2070 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRAMENP| Gene Symbol: PRAMENP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135291.1 | 6 | 2070 | |||
| GenBank mRNA | AK131325.1 | - | 6 | 2070 | ||