siRNA Details
| Gene Symbol: PRAMEF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099851.3 | NP_001093321.1 | 2 | 764 | ||
| GenBank mRNA | BC144662.1 | - | 2 | 764 | 9053193 | |
| BC146858.1 | - | 2 | 764 | 9056786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRAMEF17| Gene Symbol: PRAMEF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099851.3 | NP_001093321.1 | 2 | 764 | ||
| GenBank mRNA | BC144662.1 | - | 2 | 764 | 9053193 | |
| BC146858.1 | - | 2 | 764 | 9056786 | ||