siRNA Details
| Gene Symbol: NUPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145712.1 | NP_001139184.1 | 1 | 177 | ||
| GenBank mRNA | BC043523.1 | - | 1 | 177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUPR2| Gene Symbol: NUPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145712.1 | NP_001139184.1 | 1 | 177 | ||
| GenBank mRNA | BC043523.1 | - | 1 | 177 | ||