siRNA Details
| Gene Symbol: ZNF192P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103448.1 | 4 | 802 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF192P1| Gene Symbol: ZNF192P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103448.1 | 4 | 802 | |||