siRNA Details
| Gene Symbol: TRAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005906.1 | XP_016861395.1 | 2 | 543 | ||
| XM_017005909.1 | XP_016861398.1 | 2 | 543 | |||
| XM_017005911.1 | XP_016861400.1 | 2 | 543 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAK1| Gene Symbol: TRAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005906.1 | XP_016861395.1 | 2 | 543 | ||
| XM_017005909.1 | XP_016861398.1 | 2 | 543 | |||
| XM_017005911.1 | XP_016861400.1 | 2 | 543 | |||