siRNA Details
| Gene Symbol: LINC00871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102699.1 | 3 | 731 | |||
| NR_102700.1 | 2 | 635 | ||||
| NR_102701.1 | 6 | 831 | ||||
| NR_102702.1 | 4 | 714 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00871| Gene Symbol: LINC00871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102699.1 | 3 | 731 | |||
| NR_102700.1 | 2 | 635 | ||||
| NR_102701.1 | 6 | 831 | ||||
| NR_102702.1 | 4 | 714 | ||||