siRNA Details
| Gene Symbol: LINC00959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034125.1 | 4, 5 | 545 | |||
| GenBank mRNA | AK094118.1 | - | 4, 5 | 545 | ||
| EU250747.1 | - | 4, 5 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00959| Gene Symbol: LINC00959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034125.1 | 4, 5 | 545 | |||
| GenBank mRNA | AK094118.1 | - | 4, 5 | 545 | ||
| EU250747.1 | - | 4, 5 | 531 | |||