siRNA Details
| Gene Symbol: LOC107984138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023965.1 | XP_016879454.1 | 16 | 2306 | ||
| XM_017023966.1 | XP_016879455.1 | 16 | 2170 | |||
| XM_017023967.1 | XP_016879456.1 | 16 | 2196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107984138| Gene Symbol: LOC107984138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023965.1 | XP_016879454.1 | 16 | 2306 | ||
| XM_017023966.1 | XP_016879455.1 | 16 | 2170 | |||
| XM_017023967.1 | XP_016879456.1 | 16 | 2196 | |||