siRNA Details
| Gene Symbol: TCAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206938.1 | NP_001193867.1 | 4 | 1802 | ||
| NM_001206941.1 | NP_001193870.1 | 3 | 1173 | |||
| NM_014719.2 | NP_055534.1 | 4 | 1762 | |||
| XM_005250074.3 | XP_005250131.1 | 4 | 1802 | |||
| XM_005250075.2 | XP_005250132.1 | 4 | 1804 | |||
| XM_005250076.3 | XP_005250133.1 | 4 | 1839 | |||
| XM_005250077.2 | XP_005250134.1 | 5 | 1968 | |||
| XM_006716191.2 | XP_006716254.1 | 4 | 1768 | |||
| XM_006716192.3 | XP_006716255.1 | 4 | 1911 | |||
| XM_017012833.1 | XP_016868322.1 | 4 | 1834 | |||
| XM_017012834.1 | XP_016868323.1 | 4 | 1804 | |||
| XM_017012835.1 | XP_016868324.1 | 5 | 1968 | |||
| XM_017012836.1 | XP_016868325.1 | 4 | 1768 | |||
| GenBank mRNA | AB018281.1 | - | 4 | 1762 | ||
| AK001640.1 | - | 4 | 1838 | |||
| AK291605.1 | - | 4 | 1836 | |||
| AK299911.1 | - | 3 | 1071 | |||
| BC000609.2 | - | 4 | 1743 | 3346754 | ||

TCAF1
Genomic Map