siRNA Details
| Gene Symbol: FAM21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005751.2 | NP_001005751.1 | 10 | 974 | ||
| NM_001291398.1 | NP_001278327.1 | 10 | 974 | |||
| XM_005269806.3 | XP_005269863.2 | 10 | 922 | |||
| XM_005269807.3 | XP_005269864.2 | 10 | 922 | |||
| XM_005269808.3 | XP_005269865.2 | 10 | 922 | |||
| XM_006717831.3 | XP_006717894.2 | 10 | 922 | |||
| XM_006717832.3 | XP_006717895.2 | 10 | 922 | |||
| XM_011539786.2 | XP_011538088.1 | 10 | 922 | |||
| XM_017016227.1 | XP_016871716.1 | 10 | 922 | |||
| XM_017016228.1 | XP_016871717.1 | 10 | 922 | |||
| XM_017016229.1 | XP_016871718.1 | 10 | 922 | |||
| XM_017016230.1 | XP_016871719.1 | 9 | 787 | |||
| XM_017016231.1 | XP_016871720.1 | 10 | 922 | |||
| XM_017016232.1 | XP_016871721.1 | 10 | 922 | |||
| XM_017016233.1 | XP_016871722.1 | 7 | 497 | |||
| XM_017016234.1 | XP_016871723.1 | 11 | 1016 | |||
| XM_017016235.1 | XP_016871724.1 | 2 | 43 | |||
| GenBank mRNA | AK001686.1 | - | 7 | 625 | ||
| AK295129.1 | - | 10 | 926 | |||
| AK304116.1 | - | 10 | 910 | |||
| AK311628.1 | - | 7 | 528 | |||
| BC075815.1 | - | 7 | 539 | |||
| BC082258.1 | - | 10 | 901 | 6278433 | ||
| BC144479.1 | - | 10 | 1003 | 9053006 | ||
| DA402580.1 | - | 2 | 132 | |||
| DC403380.1 | - | 7 | 528 | |||

FAM21C
Genomic Map