siRNA Details
| Gene Symbol: CDRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282540.1 | NP_001269469.1 | 5 | 1296 | ||
| NM_006382.3 | NP_006373.2 | 5 | 1296 | |||
| GenBank mRNA | CB989113.1 | - | 3 | 600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDRT1| Gene Symbol: CDRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282540.1 | NP_001269469.1 | 5 | 1296 | ||
| NM_006382.3 | NP_006373.2 | 5 | 1296 | |||
| GenBank mRNA | CB989113.1 | - | 3 | 600 | ||