siRNA Details
| Gene Symbol: RBMY2FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002193.2 | 4 | 540 | |||
| GenBank mRNA | BC026077.2 | - | 4 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMY2FP| Gene Symbol: RBMY2FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002193.2 | 4 | 540 | |||
| GenBank mRNA | BC026077.2 | - | 4 | 542 | ||