siRNA Details
| Gene Symbol: GIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005142.2 | NP_005133.2 | 6 | 797 | ||
| XM_011544939.2 | XP_011543241.1 | 6 | 825 | |||
| GenBank mRNA | AK301295.1 | - | 6 | 909 | ||
| AK314275.1 | - | 6 | 816 | |||
| BC037958.1 | - | 6 | 797 | 5759934 | ||
| X76562.1 | - | 6 | 796 | |||

GIF
Genomic Map