siRNA Details
Gene Symbol: GOLGA8T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033933.1 | 8 | 670 | |||
GenBank mRNA | AK310526.1 | - | 8 | 670 |
Gene Symbol: GOLGA8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282490.1 | NP_001269419.1 | 8 | 648 |
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 8 | 650 | ||
XM_006720651.3 | XP_006720714.1 | 8 | 670 | |||
XM_011521948.2 | XP_011520250.1 | 8 | 978 | |||
XM_011521950.2 | XP_011520252.1 | 8 | 976 | |||
GenBank mRNA | DB054802.1 | - | 8 | 539 |
Gene Symbol: GOLGA8K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282493.1 | NP_001269422.1 | 8 | 650 |
