siRNA Details
| Gene Symbol: GOLGA8IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024074.1 | 11 | 1381 | |||
| GenBank mRNA | AK093104.1 | - | 11 | 1381 | ||
| Gene Symbol: GOLGA8H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282490.1 | NP_001269419.1 | 12 | 1178 | ||
| Gene Symbol: GOLGA8J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282472.1 | NP_001269401.1 | 12 | 1180 | ||
| XM_006720651.3 | XP_006720714.1 | 12 | 1200 | |||
| XM_011521948.2 | XP_011520250.1 | 12 | 1508 | |||
| XM_011521950.2 | XP_011520252.1 | 12 | 1506 | |||
| Gene Symbol: GOLGA8K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282493.1 | NP_001269422.1 | 12 | 1180 | ||
| Gene Symbol: GOLGA8M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282468.1 | NP_001269397.1 | 12 | 1180 | ||
| XM_005268284.4 | XP_005268341.1 | 12 | 1979 | |||
| XM_005268285.4 | XP_005268342.1 | 12 | 1979 | |||
| XM_006720654.3 | XP_006720717.1 | 12 | 1123 | |||
| XM_011521951.2 | XP_011520253.1 | 13 | 1548 | |||
| XM_011521952.2 | XP_011520254.1 | 12 | 1979 | |||
| XM_011521953.2 | XP_011520255.1 | 12 | 1979 | |||
| XM_011521954.2 | XP_011520256.1 | 12 | 1113 | |||
| GenBank mRNA | BF087405.1 | - | 4 | 452 | ||
| Gene Symbol: GOLGA8T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033933.1 | 12 | 1200 | |||

GOLGA8IP
Genomic Map