siRNA Details
| Gene Symbol: FAM74A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026802.1 | 2 | 994 | |||
| NR_110998.1 | 3 | 1078 | ||||
| GenBank mRNA | AK127145.1 | - | 2 | 856 | ||
| Gene Symbol: ATP2C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011523486.2 | XP_011521788.1 | 28 | 3810 | ||
| XM_011523487.2 | XP_011521789.1 | 18 | 2852 | |||

FAM74A4
Genomic Map