siRNA Details
Gene Symbol: AFDN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027906.1 | 2 | 1765 | |||
GenBank mRNA | AB016898.1 | - | 2 | 1678 | ||
AK091690.1 | - | 3 | 1667 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AFDN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027906.1 | 2 | 1765 | |||
GenBank mRNA | AB016898.1 | - | 2 | 1678 | ||
AK091690.1 | - | 3 | 1667 |