siRNA Details
| Gene Symbol: AFDN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027906.1 | 1 | 1032 | |||
| GenBank mRNA | AB016898.1 | - | 1 | 945 | ||
| AK091690.1 | - | 2 | 934 | |||
| AK301419.1 | - | 1 | 1024 | |||
| AK307980.1 | - | 2 | 686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AFDN-AS1| Gene Symbol: AFDN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027906.1 | 1 | 1032 | |||
| GenBank mRNA | AB016898.1 | - | 1 | 945 | ||
| AK091690.1 | - | 2 | 934 | |||
| AK301419.1 | - | 1 | 1024 | |||
| AK307980.1 | - | 2 | 686 | |||