siRNA Details
| Gene Symbol: CRLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022148.3 | NP_071431.2 | 1, 2 | 179 | ||
| NR_110830.1 | 1, 2 | 176 | ||||
| XM_011546181.2 | XP_011544483.1 | 1, 2 | 214 | |||
| XM_011546182.2 | XP_011544484.1 | 1, 2 | 218 | |||
| GenBank mRNA | AB052639.1 | - | 1, 2 | 90 | ||
| AF142570.1 | - | 1, 2 | 106 | |||
| AF338733.1 | - | 1, 2 | 90 | |||
| AK026800.1 | - | 1, 2 | 178 | |||
| AK223303.1 | - | 1, 2 | 206 | |||
| AK225583.1 | - | 1, 2 | 178 | |||

CRLF2
Genomic Map