siRNA Details
| Gene Symbol: TMEM236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098844.2 | NP_001092314.1 | 1 | 246 | ||
| XM_011519626.2 | XP_011517928.1 | 1 | 397 | |||
| XM_011519628.1 | XP_011517930.1 | 1 | 37 | |||
| GenBank mRNA | BC157856.1 | - | 1 | 246 | 9057075 | |

TMEM236
Genomic Map