siRNA Details
| Gene Symbol: TMEM236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098844.2 | NP_001092314.1 | 3 | 457 | ||
| XM_011519626.2 | XP_011517928.1 | 3 | 608 | |||
| XM_011519627.2 | XP_011517929.1 | 3 | 334 | |||
| XM_011519628.1 | XP_011517930.1 | 2 | 175 | |||
| XM_011519629.1 | XP_011517931.1 | 2 | 223 | |||
| XM_017016574.1 | XP_016872063.1 | 3 | 1183 | |||
| GenBank mRNA | AK300456.1 | - | 2 | 200 | ||
| BC157856.1 | - | 3 | 457 | 9057075 | ||

TMEM236
Genomic Map