siRNA Details
| Gene Symbol: PHLDB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198850.3 | NP_942147.3 | 10, 11 | 1500 | ||
| XM_005259169.3 | XP_005259226.1 | 10, 11 | 1568 | |||
| XM_005259170.3 | XP_005259227.1 | 10, 11 | 1568 | |||
| XM_005259171.3 | XP_005259228.1 | 10, 11 | 1568 | |||
| XM_011527236.2 | XP_011525538.1 | 10, 11 | 1672 | |||
| GenBank mRNA | BC157852.1 | - | 4, 5 | 484 | ||
| BC171853.1 | - | 4, 5 | 493 | |||
| BG403419.1 | - | 4, 5 | 351 | |||

PHLDB3
Genomic Map