siRNA Details
| Gene Symbol: PHLDB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198850.3 | NP_942147.3 | 15 | 2022 | ||
| XM_005259169.3 | XP_005259226.1 | 15 | 2090 | |||
| XM_005259170.3 | XP_005259227.1 | 15 | 1988 | |||
| XM_011527236.2 | XP_011525538.1 | 15 | 2194 | |||
| GenBank mRNA | BC007947.2 | - | 4 | 377 | ||
| BC157852.1 | - | 9 | 904 | |||
| BC171853.1 | - | 9 | 913 | |||

PHLDB3
Genomic Map