siRNA Details
| Gene Symbol: GOLGA6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164404.1 | NP_001157876.1 | 3 | 300 | ||
| XM_017022519.1 | XP_016878008.1 | 3 | 454 | |||
| Gene Symbol: GOLGA6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF263742.1 | - | 3 | 300 | ||
| Gene Symbol: GOLGA6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145224.1 | NP_001138696.1 | 3 | 294 | ||

GOLGA6C
Genomic Map