siRNA Details
| Gene Symbol: GOLGA6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145224.1 | NP_001138696.1 | 7 | 530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOLGA6D| Gene Symbol: GOLGA6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145224.1 | NP_001138696.1 | 7 | 530 | ||