siRNA Details
| Gene Symbol: TMEM183B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079809.1 | NP_001073277.1 | 1 | 817 | ||
| GenBank mRNA | AK297222.1 | - | 6 | 737 | ||
| Gene Symbol: TMEM183A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322955.1 | NP_001309884.1 | 6 | 855 | ||
| NM_001322956.1 | NP_001309885.1 | 6 | 855 | |||
| NM_001322957.1 | NP_001309886.1 | 6 | 858 | |||
| NM_001322958.1 | NP_001309887.1 | 5 | 768 | |||
| NM_001322959.1 | NP_001309888.1 | 5 | 765 | |||
| NM_138391.4 | NP_612400.3 | 6 | 819 | |||
| NR_136530.1 | 7 | 1022 | ||||
| NR_136531.1 | 6 | 835 | ||||
| NR_136532.1 | 6 | 838 | ||||
| NR_136533.1 | 7 | 1019 | ||||
| NR_136534.1 | 7 | 1019 | ||||
| GenBank mRNA | AF070537.1 | - | 6 | 836 | ||
| AK291426.1 | - | 6 | 833 | |||
| AK312985.1 | - | 6 | 833 | |||
| BC013073.1 | - | 1 | 819 | 4151716 | ||
| BC038952.2 | - | 1 | 871 | 5740484 | ||
| BC067775.1 | - | 7 | 993 | 4817183 | ||
| BC150647.1 | - | 6 | 801 | 9057018 | ||

TMEM183B
Genomic Map