siRNA Details
| Gene Symbol: TMEM217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286401.1 | NP_001273330.1 | 3 | 1544 | ||
| NM_145316.3 | NP_660359.2 | 4 | 1111 | |||
| NR_104442.1 | 3 | 1013 | ||||
| NR_104443.1 | 2 | 928 | ||||
| XM_011514367.2 | XP_011512669.1 | 4 | 1038 | |||
| XM_011514369.2 | XP_011512671.1 | 2 | 439 | |||
| XM_011514370.2 | XP_011512672.1 | 2 | 429 | |||
| GenBank mRNA | AK098666.1 | - | 4 | 1070 | ||
| BC026012.1 | - | 3 | 983 | 5269554 | ||
| BC029657.1 | - | 2 | 409 | 5167418 | ||

TMEM217
Genomic Map