siRNA Details
Gene Symbol: UTP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014503.2 | NP_055318.2 | 6, 7 | 761 | ||
GenBank mRNA | AJ006778.1 | - | 6, 7 | 749 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UTP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014503.2 | NP_055318.2 | 6, 7 | 761 | ||
GenBank mRNA | AJ006778.1 | - | 6, 7 | 749 |