siRNA Details
| Gene Symbol: ARPC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005720.3 | NP_005711.1 | 4, 5 | 509 | ||
| XM_006715825.1 | XP_006715888.1 | 4, 5 | 520 | |||
| XM_006715826.1 | XP_006715889.1 | 4, 5 | 504 | |||
| GenBank mRNA | AF006084.1 | - | 4, 5 | 469 | ||
| AK314818.1 | - | 4, 5 | 454 | |||
| AU143525.1 | - | 4, 5 | 509 | |||
| AY007111.1 | - | 2, 3 | 339 | |||
| BC002562.2 | - | 4, 5 | 442 | 3138319 | ||
| BC002988.1 | - | 4, 5 | 478 | 2821335 | ||
| BC007555.1 | - | 4, 5 | 440 | 2967684 | ||

ARPC1B
Genomic Map