siRNA Details
| Gene Symbol: NUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138459.3 | NP_612468.1 | 4, 5 | 1018 | ||
| GenBank mRNA | AK125119.1 | - | 4, 5 | 980 | ||
| AK126335.1 | - | 5, 6 | 732 | |||
| AK130093.1 | - | 3, 4 | 335 | |||
| BC013026.2 | - | 4, 5 | 965 | 3531630 | ||
| BC063794.1 | - | 4, 5 | 972 | 6500832 | ||
| BC066910.1 | - | 4, 5 | 1012 | 4830860 | ||
| BC110325.1 | - | 4, 5 | 974 | 5553827 | ||
| BC150654.1 | - | 4, 5 | 985 | 9057025 | ||
| BC150655.1 | - | 4, 5 | 985 | 9057026 | ||

NUS1
Genomic Map