siRNA Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000167.5 | NP_000158.1 | 5 | 559 | ||
| NM_001128127.2 | NP_001121599.1 | 5 | 559 | |||
| NM_001205019.1 | NP_001191948.1 | 5 | 559 | |||
| NM_203391.3 | NP_976325.1 | 5 | 559 | |||
| XM_005274488.4 | XP_005274545.1 | 3 | 329 | |||
| XM_006724483.2 | XP_006724546.1 | 6 | 596 | |||
| XM_006724484.2 | XP_006724547.1 | 6 | 595 | |||
| XM_006724485.2 | XP_006724548.1 | 4 | 404 | |||
| XM_006724486.3 | XP_006724549.1 | 3 | 389 | |||
| XM_011545491.2 | XP_011543793.1 | 6 | 597 | |||
| XM_011545492.1 | XP_011543794.1 | 6 | 596 | |||
| XM_011545493.2 | XP_011543795.1 | 4 | 279 | |||
| XM_011545494.2 | XP_011543796.1 | 4 | 224 | |||
| XM_017029409.1 | XP_016884898.1 | 6 | 594 | |||
| XM_017029410.1 | XP_016884899.1 | 3 | 373 | |||
| XM_017029411.1 | XP_016884900.1 | 4 | 406 | |||
| XM_017029412.1 | XP_016884901.1 | 3 | 327 | |||
| GenBank mRNA | AK294304.1 | - | 4 | 315 | ||
| AK294938.1 | - | 6 | 594 | |||
| AK308238.1 | - | 3 | 238 | |||
| AK313215.1 | - | 5 | 495 | |||
| BC037549.1 | - | 5 | 416 | 5213716 | ||
| BC042421.1 | - | 5 | 560 | 4820945 | ||
| BC071595.1 | - | 5 | 560 | 5276056 | ||
| BM906503.1 | - | 506 | ||||
| BX647604.1 | - | 5 | 380 | |||
| L13943.1 | - | 5 | 446 | |||
| X68285.1 | - | 3 | 152 | |||
| Gene Symbol: GK3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026575.1 | 1 | 758 | |||
| GenBank mRNA | AK225740.1 | - | 1 | 409 | ||
| AK292282.1 | - | 1 | 758 | |||
| BC066960.1 | - | 1 | 414 | |||

GK
Genomic Map