siRNA Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000167.5 | NP_000158.1 | 2 | 292 | ||
| NM_001128127.2 | NP_001121599.1 | 2 | 292 | |||
| NM_001205019.1 | NP_001191948.1 | 2 | 292 | |||
| NM_203391.3 | NP_976325.1 | 2 | 292 | |||
| XM_006724483.2 | XP_006724546.1 | 2 | 245 | |||
| XM_006724484.2 | XP_006724547.1 | 2 | 244 | |||
| XM_006724485.2 | XP_006724548.1 | 2 | 244 | |||
| XM_011545491.2 | XP_011543793.1 | 2 | 246 | |||
| XM_011545492.1 | XP_011543794.1 | 2 | 245 | |||
| XM_017029409.1 | XP_016884898.1 | 2 | 162 | |||
| XM_017029411.1 | XP_016884900.1 | 2 | 246 | |||
| GenBank mRNA | AK294304.1 | - | 2 | 155 | ||
| AK294938.1 | - | 2 | 162 | |||
| AK313215.1 | - | 2 | 228 | |||
| BC037549.1 | - | 2 | 149 | 5213716 | ||
| BC042421.1 | - | 2 | 293 | 4820945 | ||
| BC071595.1 | - | 2 | 293 | 5276056 | ||
| BM906503.1 | - | 239 | ||||
| BX647604.1 | - | 2 | 153 | |||
| L13943.1 | - | 2 | 179 | |||

GK
Genomic Map