siRNA Details
| Gene Symbol: AREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001657.3 | NP_001648.1 | 2 | 508 | ||
| GenBank mRNA | BC009799.1 | - | 2 | 474 | 4277616 | |
| BC146953.1 | - | 2 | 315 | 9056881 | ||
| BC146967.1 | - | 2 | 315 | 9056895 | ||
| BT019866.1 | - | 2 | 260 | |||
| M30704.1 | - | 2 | 469 | |||

AREG
Genomic Map