siRNA Details
| Gene Symbol: RNF208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031297.4 | NP_112587.2 | 1 | 457 | ||
| GenBank mRNA | AF416715.1 | - | 1 | 138 | ||
| AL136540.1 | - | 1 | 140 | |||
| BC016958.1 | - | 1 | 146 | 4151712 | ||
| DN995061.1 | - | 1 | 220 | |||

RNF208
Genomic Map