siRNA Details
| Gene Symbol: LOC101928424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135053.1 | 2 | 579 | |||
| GenBank mRNA | BX104024.1 | - | 2 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928424| Gene Symbol: LOC101928424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135053.1 | 2 | 579 | |||
| GenBank mRNA | BX104024.1 | - | 2 | 564 | ||