siRNA Details
| Gene Symbol: NBPF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095459.1 | - | 3 | 409 | ||
| Gene Symbol: NBPF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR936830.1 | - | 3 | 1921 | ||

NBPF14
Genomic Map