siRNA Details
| Gene Symbol: RHOXF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032498.2 | NP_115887.1 | 3, 4 | 735 | ||
| XM_005262484.3 | XP_005262541.1 | 3, 4 | 749 | |||
| GenBank mRNA | AF317219.1 | - | 3, 4 | 675 | ||
| AK058125.1 | - | 3, 4 | 628 | |||
| AL590526.1 | - | 3, 4 | 735 | |||
| BC021719.2 | - | 3, 4 | 630 | 5269333 | ||
| Gene Symbol: RHOXF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099685.1 | NP_001093155.1 | 3, 4 | 735 | ||
| GenBank mRNA | AL590524.1 | - | 3, 4 | 631 | ||

RHOXF2
Genomic Map