siRNA Details
| Gene Symbol: EYS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142800.1 | NP_001136272.1 | 43 | 10370 | ||
| NM_001292009.1 | NP_001278938.1 | 44 | 10433 | |||
| GenBank mRNA | FJ416331.1 | - | 43 | 10378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EYS| Gene Symbol: EYS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142800.1 | NP_001136272.1 | 43 | 10370 | ||
| NM_001292009.1 | NP_001278938.1 | 44 | 10433 | |||
| GenBank mRNA | FJ416331.1 | - | 43 | 10378 | ||