siRNA Details
| Gene Symbol: LOC105378397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134317.1 | 5 | 397 | |||
| GenBank mRNA | BX092644.1 | - | 5 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378397| Gene Symbol: LOC105378397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134317.1 | 5 | 397 | |||
| GenBank mRNA | BX092644.1 | - | 5 | 397 | ||