siRNA Details
| Gene Symbol: FAM133DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034169.1 | 1 | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM133DP| Gene Symbol: FAM133DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034169.1 | 1 | 696 | |||